Learn More
Sino Biological Recombinant Rhesus IL-13 Protein (His Tag) 100 µg
Catalog No. 5000656843
Encompass_Preferred
A DNA sequence encoding the Rhesus IL-13 (Q864V6) (Leu11-Asn132) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-006-56843 Supplier Sino Biological Supplier No. 91102K08H100
A DNA sequence encoding the Rhesus IL-13 (Q864V6) (Leu11-Asn132) was expressed with a polyhistidine tag at the C-terminus.