Learn More
Sino Biological Protein RSV RSV-G 100UG
Catalog No. 5001073928
Encompass_Preferred
A DNA sequence encoding the Human respiratory syncytial virus (RSVA) (HRSV/A/USA/TX-HOU-R06687/2021) glycoprotein G / RSV-G (XHB13219 1) (Ser64-Lys321) was expressed fused with a polyhistidine tag at the C-terminus
Catalog No. 50-010-73928 Supplier Sino Biological Supplier No. 41138V08H100
A DNA sequence encoding the Human respiratory syncytial virus (RSVA) (HRSV/A/USA/TX-HOU-R06687/2021) glycoprotein G / RSV-G (XHB13219 1) (Ser64-Lys321) was expressed fused with a polyhistidine tag at the C-terminus