Learn More
Sino Biological Protein RSV RSV-G 100UG
Catalog No. 5001073849
Encompass_Preferred
A DNA sequence encoding the Human respiratory syncytial virus (RSVA) (RSV/Human/USA/MA-Broad MGB-13665/2022) glycoprotein G / RSV-G (WAO97722 1) (Ser64-Lys321) was expressed with a polyhistidine tag at the C-terminus
Catalog No. 50-010-73849 Supplier Sino Biological Supplier No. 41144V08H100
A DNA sequence encoding the Human respiratory syncytial virus (RSVA) (RSV/Human/USA/MA-Broad MGB-13665/2022) glycoprotein G / RSV-G (WAO97722 1) (Ser64-Lys321) was expressed with a polyhistidine tag at the C-terminus