Learn More
Sino Biological Protein RSV RSV-G 100UG
Catalog No. 5001074070
Encompass_Preferred
A DNA sequence encoding the Human respiratory syncytial virus (RSVB) (HRSV/B/USA/TX-HOU-IP0187A/2021) G (XHB13252 1) (Ser64-Thr310) was expressed with a polyhistidine tag at the C-terminus
Catalog No. 50-010-74070 Supplier Sino Biological Supplier No. 41132V08H100
A DNA sequence encoding the Human respiratory syncytial virus (RSVB) (HRSV/B/USA/TX-HOU-IP0187A/2021) G (XHB13252 1) (Ser64-Thr310) was expressed with a polyhistidine tag at the C-terminus