Learn More
Sino Biological Protein RVFV-NP Biotin 20UG
Catalog No. 5001074120
Encompass_Preferred
A DNA sequence encoding the Rift Valley fever virus (TAN/Dod-002/07) RVFV-NP (P21700) (Met1-Ala245) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed
Catalog No. 50-010-74120 Supplier Sino Biological Supplier No. 41190V49EB20
A DNA sequence encoding the Rift Valley fever virus (TAN/Dod-002/07) RVFV-NP (P21700) (Met1-Ala245) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed