Learn More
BioVendor, LLC SARS-CoV Nucleocapsid Protein E. coli, 0.1 MG
SDP
Supplier:  BioVendor, LLC RD972052100
Encompass_Preferred
TypeRecombinant proteinDescriptionTotal 437 AA. MW: 47.8 kDa (calculated). UniProtKB acc.no. P59595. N-Terminal His-tag 16 AA (highlighted).Amino Acid SequenceMRGSHHHHHHGMASHMSDNGPQSNQRSAPRITFGGPTDSTDNNQNGGRNGARPKQRRPQGLPNNTASWFTALTQHGKEELRFPRGQGVPINTNSGPDDQIGYYRRATRRVRGGDGKMKELSPRWYFYYLGTGPEASLPYGANKEGIVWVATEGALNTPKDHIGTRNPNNNAATVLQLPQGTTLPKGFYAEGSRGGSQASSRSSSRSRGNSRNSTPGSSRGNSPARMASGGGETALALLLLDRLNQLESKVSGKGQQQQGQTVTKKSAAEASKKPRQKRTATKQYNVTQAFGRRGPEQTQGNFGDQDLIRQGTDYKHWPQIAQFAPSASAFFGMSRIGMEVTPSGTWLTYHGAIKLDDKDPQFKDNVILLNKHIDAYKTFPPTEPKKDKKKKTDEAQPLPQRQKKQPTVTLLPAADMDDFSRQLQNSMSGASADSTQASourceE. coliPurity>95%SDS-PAGE Gel12% SDS-PAGE separation of SARS-CoV Nucleocapsid protein1. M.W. marker – 14, 21, 31, 45, 66, 97 kDa2. reduced and heated sample, 5μg/lane3. non-reduced and non-heated sample, 5μg/laneEndotoxin< 0.1 EU/μgApplicationsWestern blotting
Catalog No. 50-894-447