Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) NSP10-His Recombinant Protein, Biotinylated 100 µg
Catalog No. 5000656576
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) NSP10 (YP_009725306.1) (Ala1-Gln139) was expressed with an initial Met at the N-terminus and a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-006-56576 Supplier Sino Biological Supplier No. 40599V07EB100
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) NSP10 (YP_009725306.1) (Ala1-Gln139) was expressed with an initial Met at the N-terminus and a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.