Learn More
Sino Biological Protein SARS-CoV-2 Spike 100UG
Catalog No. 5001073915
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (BA 3 2) Spike S1 S2 trimer (YP 009724390 1) (Met1-Gln1208) with a C-terminal polyhistidine tag was expressed
Catalog No. 50-010-73915 Supplier Sino Biological Supplier No. 40589V08H75100
A DNA sequence encoding the SARS-CoV-2 (BA 3 2) Spike S1 S2 trimer (YP 009724390 1) (Met1-Gln1208) with a C-terminal polyhistidine tag was expressed