Learn More
Sino Biological Protein SARS-CoV-2 Spike 100UG
Catalog No. 5001074081
Encompass_Preferred
A DNA sequence encoding the SARS-COV-2 BA 3 2 (Omicron) Spike RBD (YP 009724390 1 with mutation I326V G339Y A348P K356T S371F S373P S375F R403K D405N R408S K417N A435S N440R V445A G446D L452W N460K S477N T478N E484K G496S Q498R N501Y K529N) (Arg319-Lys529) was expressed with a polyhistidine tag at the C-terminus The mutations were identified in the SARS-CoV-2 variant (known as variant BA 3 2)
Catalog No. 50-010-74081 Supplier Sino Biological Supplier No. 40592V08H164100
A DNA sequence encoding the SARS-COV-2 BA 3 2 (Omicron) Spike RBD (YP 009724390 1 with mutation I326V G339Y A348P K356T S371F S373P S375F R403K D405N R408S K417N A435S N440R V445A G446D L452W N460K S477N T478N E484K G496S Q498R N501Y K529N) (Arg319-Lys529) was expressed with a polyhistidine tag at the C-terminus The mutations were identified in the SARS-CoV-2 variant (known as variant BA 3 2)