Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Protein Saver 903 Card
SDP

Catalog No. 22445975 Shop All Mopec Products
Change view
Click to view available options
Dimensions (L x W):
3.5 x 2.67 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-445-975 3.5 x 2.67 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-445-975 Supplier Mopec Supplier No. FS259
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reliable sample collection and storage solution that helps preserve protein integrity during transport and Long-term storage.

  • Each circle holds 75 to 80 μL of sample.
  • Printed on both front and back.
  • Compatible with various types of fingerprinting ink.
  • Enables easy application of biological samples.
  • Supports rapid sample drying.
  • Helps maintain sample stability at ambient temperatures.
  • Features 5 circles, each measuring 0.5 in.
  • Made with High-quality absorbent paper

Specifications

Dimensions (L x W) 3.5 x 2.67 in.
Product Type Protein Saver Card
Color White with Black Print
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.