Learn More
Sino Biological Protein SerpinA1 ET-FREE 100UG
Catalog No. 5001073967
Encompass_Preferred
A DNA sequence encoding the human SerpinA1 (NP 000286 3) pre-protein (Met 1-Lys 418) was expressed with a C-terminal polyhistidine tag
Catalog No. 50-010-73967 Supplier Sino Biological Supplier No. 10306H08HUE100
A DNA sequence encoding the human SerpinA1 (NP 000286 3) pre-protein (Met 1-Lys 418) was expressed with a C-terminal polyhistidine tag