Learn More
Sino Biological Protein SerpinE1 ET-FREE 1MG
Catalog No. 5001073682
Encompass_Preferred
A DNA sequence encoding the human SerpinE1 precursor (NP 000593 1) (Met 1-Pro 402) was expressed with a C-terminal polyhistidine tag
Catalog No. 50-010-73682 Supplier Sino Biological Supplier No. 10296H08HUE1
A DNA sequence encoding the human SerpinE1 precursor (NP 000593 1) (Met 1-Pro 402) was expressed with a C-terminal polyhistidine tag