Learn More
Invent Biotechnologies Inc Minute™ Protein Solubilization Reagent for MS (2 ml)
Catalog No. NC1740002
Encompass
This item has a minimum qty of 4 per supplier requirements.
This reagent is formulated for dissolving protein pellets using protein kits from Invent Biotechnologies (Cat# Nos. SM-005, SM-005-P, CP-011, NE-013, YM-017, MM-018 and AF-023) and other companies. Solubilized proteins are denatured due to the presence of SDS and can be used for trypsin digestion and for subsequent mass spectrometry analysis.
Catalog No. NC1740002 Supplier Invent Biotechnologies Inc Supplier No. WA011
This item has a minimum qty of 4 per supplier requirements.
This reagent is formulated for dissolving protein pellets using protein kits from Invent Biotechnologies (Cat# Nos. SM-005, SM-005-P, CP-011, NE-013, YM-017, MM-018 and AF-023) and other companies. Solubilized proteins are denatured due to the presence of SDS and can be used for trypsin digestion and for subsequent mass spectrometry analysis.