Learn More
Protein Standard Mix 15 - 600 kDa for size exclusion chromatography, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| CAS | 26628-22-8 |
| Physical Form | Neat |
| Technique | Gas Chromatography |
| Type | Organic Standard |
| Solvent or Matrix | Neat |
| Industry Type | Laboratory Use |
| Quantity | 6 x 30 mg |
| Grade | Reference Material |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.