Learn More
Protein Standard Mix 15 - 600 kDa for size exclusion chromatography, MilliporeSigma™ Supelco™
Catalog No. p-7187556 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
30 mg
6 x 30 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3779 30 mg
11-100-2597 6 x 30 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3779 Supplier MilliporeSigma Supelco Supplier No. 6938530MG
Only null left
Add to Cart
Add to Cart
The protein standard mix is a calibration standard to test and monitor performance of size exclusion chromatography (SEC) columns. It is a lyophilized mixture of molecular weight markers ranging from 15 kDa to 600 kDa.
Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY
Specifications
Physical Form Neat
Type Organic Standard
Quantity 30 mg