Learn More
BioVendor, LLC PROTEIN Stanniocalcin 1 Human HEK293, 0.1 MG
SDP
Supplier:  BioVendor, LLC RD172095100
Encompass
TypeRecombinant proteinDescriptionTotal 240 AA. Mw: 25.9 kDa (calculated). C-Terminal Flag-tag, 10AA (highlighted). The AA sequence is identical to Swiss-Prot- P52823.Amino Acid SequenceTHEAEQNDSVSPRKSRVAAQNSAEVVRCLNSALQVGCGAFACLENSTCDTDGMYDICKSFLYSAAKFDTQGKAFVKESLKCIANGVTSKVFLAIRRCSTFQRMIAEVQEECYSKLNVCSIAKRNPEAITEVVQLPNHFSNRYYNRLVRSLLECDEDTVSTIRDSLMEKIGPNMASLFHILQTDHCAQTHPRADFNRRRTNEPQKLKVLLRNLRGEEDSPSHIKRTSHESAASDYKDDDDK.His 2 to Glu 5 were confirmed by N-terminal sequencingSourceHEK293Purity>95% Endotoxin< 0.1 EU/μgApplicationsWestern blotting, ELISA, Cell culture and/or animal studies
Catalog No. 50-894-395