Learn More
Cell Signaling Technology ChIP-Grade Protein G Agarose Beads, 1 mL
SDP
Catalog No. 50195285
Encompass
ChIP-Grade Protein G Agarose Beads are an affinity matrix designed for the small-scale isolation of immunocomplexes in chromatin immunoprecipitation (ChIP) assays. A truncated form of recombinant Protein G is covalently coupled to agarose, providing high affinity for IgG subclasses from species including human, rabbit, mouse, rat, and sheep. The beads are provided in a storage buffer containing blocking agents to minimize non-specific binding.
  • Features truncated recombinant Protein G for high affinity across multiple species
  • Includes 500 μg/mL BSA and 200 μg/mL sonicated salmon sperm DNA to block non-specific binding
  • Supplied in a storage buffer with 10 mM Tris-HCl (pH 7.5), 1 mM EDTA, and 0.05% sodium azide
  • Not compatible with ChIP-seq due to interference from sonicated salmon sperm DNA
Catalog No. 50-195-285 Supplier Cell Signaling Technology Supplier No. 9007S
ChIP-Grade Protein G Agarose Beads are an affinity matrix designed for the small-scale isolation of immunocomplexes in chromatin immunoprecipitation (ChIP) assays. A truncated form of recombinant Protein G is covalently coupled to agarose, providing high affinity for IgG subclasses from species including human, rabbit, mouse, rat, and sheep. The beads are provided in a storage buffer containing blocking agents to minimize non-specific binding.
  • Features truncated recombinant Protein G for high affinity across multiple species
  • Includes 500 μg/mL BSA and 200 μg/mL sonicated salmon sperm DNA to block non-specific binding
  • Supplied in a storage buffer with 10 mM Tris-HCl (pH 7.5), 1 mM EDTA, and 0.05% sodium azide
  • Not compatible with ChIP-seq due to interference from sonicated salmon sperm DNA