Learn More
Sino Biological Protein TACSTD2 AF647 100UG
Catalog No. 5001073661
Encompass_Preferred
A DNA sequence encoding the human TROP-2 (NP 002344 2) (Met 1-Thr 274) was expressed with a C-terminal polyhistidine tag The protein is site-specifically conjugated with AF 647 (Excitation Max 655 nm Emission Max 680 nm)
Catalog No. 50-010-73661 Supplier Sino Biological Supplier No. 10428H86HSG100
A DNA sequence encoding the human TROP-2 (NP 002344 2) (Met 1-Thr 274) was expressed with a C-terminal polyhistidine tag The protein is site-specifically conjugated with AF 647 (Excitation Max 655 nm Emission Max 680 nm)