Learn More
Crescent Chemical Co Inc PROTEIN TEST MIX 4 FOR SDS PG
SDP
Catalog No. 50248342
Encompass_Preferred
Protein Test Mixture 4 for SDS PAGE 4 Dalton-marker proteins, high molecular weight, for SDS PAGE Molecular weight markers for SDS gel electrophoresis. The bovine albumin (BSA) contains monomers and oligomers.Reconstitute with SDS sample buffer (125 mM Tris-HCl pH 6.8, 2 % SDS, 15 %
Catalog No. 50-248-342 Supplier Crescent Chemical Co Inc Supplier No. SE39208.01
Protein Test Mixture 4 for SDS PAGE 4 Dalton-marker proteins, high molecular weight, for SDS PAGE Molecular weight markers for SDS gel electrophoresis. The bovine albumin (BSA) contains monomers and oligomers.Reconstitute with SDS sample buffer (125 mM Tris-HCl pH 6.8, 2 % SDS, 15 %