Learn More
Crescent Chemical Co Inc PROTEIN TEST MIX 5 FOR SDS PG
SDP
Catalog No. 50248343
Encompass_Preferred
Protein Test Mixture 5 for SDS PAGE Dalton-marker proteins, low molecular weight, for SDS PAGE Molecular weight markers for SDS gel electrophoresis.Reconstitute with SDS sample buffer (125 mM Tris-HCl pH 6.8, 2 % SDS, 15 % glycerol, 10 mM DTT, 0.025 % bromophenol blue, 0.025 % Orange
Catalog No. 50-248-343 Supplier Crescent Chemical Co Inc Supplier No. SE39209.01
Add to Cart
Add to Cart
Protein Test Mixture 5 for SDS PAGE Dalton-marker proteins, low molecular weight, for SDS PAGE Molecular weight markers for SDS gel electrophoresis.Reconstitute with SDS sample buffer (125 mM Tris-HCl pH 6.8, 2 % SDS, 15 % glycerol, 10 mM DTT, 0.025 % bromophenol blue, 0.025 % Orange