Learn More
Sino Biological Protein TFRC ET-FREE 100UG
Catalog No. 5001073984
Encompass_Preferred
A DNA sequence encoding the human TFRC (CAA25527 1) (Cys89-Phe760) was expressed with the Fc region of human IgG1 at the N-terminus
Catalog No. 50-010-73984 Supplier Sino Biological Supplier No. 11020H01HUE100
A DNA sequence encoding the human TFRC (CAA25527 1) (Cys89-Phe760) was expressed with the Fc region of human IgG1 at the N-terminus