Learn More
Sino Biological Protein TGFB1 ET-FREE 100UG
Catalog No. 5001074044
Encompass_Preferred
A DNA sequence encoding the Human Rhesus Cynomolgus Canine TGFB1 (NP 000651 3) (Ala279-Ser390) was expressed with the Fc region of human IgG1 at the C-terminus
Catalog No. 50-010-74044 Supplier Sino Biological Supplier No. 10804H02H1UE100
A DNA sequence encoding the Human Rhesus Cynomolgus Canine TGFB1 (NP 000651 3) (Ala279-Ser390) was expressed with the Fc region of human IgG1 at the C-terminus