Learn More
Sino Biological Protein TNFRSF1A Biotin 20UG
Catalog No. 5001074046
Encompass_Preferred
A DNA sequence encoding the Human TNFR1/CD120a (NP 001056 1) (Met1-Thr211) was fused with an AVI tag at the C-terminus The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed
Catalog No. 50-010-74046 Supplier Sino Biological Supplier No. 10872H17HB20
A DNA sequence encoding the Human TNFR1/CD120a (NP 001056 1) (Met1-Thr211) was fused with an AVI tag at the C-terminus The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed