Learn More
Sino Biological Protein TNFSF13B ET-FREE 100UG
Catalog No. 5001073861
Encompass_Preferred
A DNA sequence encoding the Human TNFSF13B (NP 006564 1) (Glu140-Leu285) was expressed with a polyhistidine tag at the N-terminus This construct has a trimer design
Catalog No. 50-010-73861 Supplier Sino Biological Supplier No. 10056H07HUE100
A DNA sequence encoding the Human TNFSF13B (NP 006564 1) (Glu140-Leu285) was expressed with a polyhistidine tag at the N-terminus This construct has a trimer design