Learn More
Sino Biological Protein TNFSF13B ET-FREE 1MG
Catalog No. 5001073752
Encompass_Preferred
A DNA sequence encoding the soluble form of human BAFF (Q9Y275-1) (Ala 134-Leu 285) was fused with the Fc region of human IgG1 at the N-terminus
Catalog No. 50-010-73752 Supplier Sino Biological Supplier No. 10056H01HUE1
A DNA sequence encoding the soluble form of human BAFF (Q9Y275-1) (Ala 134-Leu 285) was fused with the Fc region of human IgG1 at the N-terminus