Learn More
Invitrogen™ eBioscience™ Protein Transport Inhibitor Cocktail (500X)
Catalog No. 509309
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
4 x 100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-930-9 100 μL
50-112-9032 4 x 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-930-9 Supplier Invitrogen™ Supplier No. 00498093
  • $55.00
    $51.65 / Each of 1
    Save $3.35
    6% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart
Suitable for use in assays in which prevention of protein transport to the extracellular space is desired.
  • A cocktail of Brefeldin A and Monensin
  • Suitable for use in assays in which prevention of protein transport to the extracellular space is desired
  • It has been specifically formulated for use as a negative control for the Cell Stimulation Cocktail (plus protein transport inhibitors) (500X)
  • Addition of this cocktail to cells in culture inhibits the intracellular protein secretory/transport pathway resulting in the accumulation of secreted proteins in the lumen of the endoplasmic reticulum and in the Golgi apparatus
  • This accumulation can then be detected by intracellular staining and flow cytometric analysis or western blotting
TRUSTED_SUSTAINABILITY
Specifications
Product Line eBioscience
Type Protein Transport Inhibitor Cocktail
Quantity 100 μL
For Research Use Only. Not for use in diagnostic procedures.