Learn More
Athens Research And Technology Inc PROTEINASE 3 NEUTROPHIL 100UG
Catalog No. 50489900
Encompass_Preferred
Proteinase 3, Human Neutrophil: 100 UG; Frozen from 50 mM MES, pH 4.5 with 700 mM NaCl; MW 29,000; EC=1.0; Purity: > =95% by SDS-PAGE. Native; Purified from whole blood in the Athens Research & Technology laboratory. Serine Proteinase, Molecular Biology Reagent
Catalog No. 50-489-900 Supplier Athens Research And Technology Inc Supplier No. 1614161820100UG
Proteinase 3, Human Neutrophil: 100 UG; Frozen from 50 mM MES, pH 4.5 with 700 mM NaCl; MW 29,000; EC=1.0; Purity: > =95% by SDS-PAGE. Native; Purified from whole blood in the Athens Research & Technology laboratory. Serine Proteinase, Molecular Biology Reagent