Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Proteinase, Aspergillus oryzae | 9001-92-7 | 500 MG
SDP

Catalog No. 5000410104
Encompass_Preferred

Proteinase, Aspergillus oryzae is a serine protease, optimally active at pH 10.5, that hydrolyzes peptide bonds and efficiently degrades substrates like casein. Its activity is inhibited by DFP and potato inhibitor, and it catalyzes proteolysis via serine residues. This enzyme is notable for its high yield and cost-effective production, finding applications in food processing, detergents, and the leather industry.

  • Serine protease
  • Optimal pH 10.5
  • Efficiently degrades casein, poly-L-glutamic acid, and poly-L-lysine
  • Activity irreversibly inhibited by diisopropylfluorophosphate (DFP) and potato inhibitor
  • Catalyzes proteolysis via serine residues in its active site
  • High yield in solid-state fermentation and cost-effective production
  • Applications in food processing (e.g., soy sauce fermentation)
  • Applications in detergents
  • Applications in leather industries

Catalog No. 50-004-10104 Supplier Medchemexpress LLC Supplier No. HYB2228500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Proteinase, Aspergillus oryzae is a serine protease, optimally active at pH 10.5, that hydrolyzes peptide bonds and efficiently degrades substrates like casein. Its activity is inhibited by DFP and potato inhibitor, and it catalyzes proteolysis via serine residues. This enzyme is notable for its high yield and cost-effective production, finding applications in food processing, detergents, and the leather industry.

  • Serine protease
  • Optimal pH 10.5
  • Efficiently degrades casein, poly-L-glutamic acid, and poly-L-lysine
  • Activity irreversibly inhibited by diisopropylfluorophosphate (DFP) and potato inhibitor
  • Catalyzes proteolysis via serine residues in its active site
  • High yield in solid-state fermentation and cost-effective production
  • Applications in food processing (e.g., soy sauce fermentation)
  • Applications in detergents
  • Applications in leather industries

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.