Learn More
Medchemexpress LLC Proteinase-inhibiting protein Bowman-Birk | 37330-34-0 | MFCD00132532 | 70-90% | approximately 8 kDa | 25 MG
SDP
Catalog No. 5000398807
Encompass_Preferred
The Bowman-Birk inhibitor is a highly cross-linked protein with seven disulfide bridges, featuring spatially distinct domains. It plays a significant role as a plant protease inhibitor.
  • Specifically inhibits both trypsin and chymotrypsin
  • Exhibits anticarcinogenic properties
Catalog No. 50-003-98807 Supplier Medchemexpress LLC Supplier No. 25MGHYP3026
The Bowman-Birk inhibitor is a highly cross-linked protein with seven disulfide bridges, featuring spatially distinct domains. It plays a significant role as a plant protease inhibitor.
  • Specifically inhibits both trypsin and chymotrypsin
  • Exhibits anticarcinogenic properties