Learn More
Alkali Scientific Proteinase K, 1 G
SDP
Catalog No. 5000659998
Encompass_Preferred
Proteinase K is a highly active protease enzyme used primarily in DNA extraction protocols. This enzyme efficiently digests proteins and helps in the removal of contaminants that may inhibit downstream applications. It is particularly useful in isolating high-quality DNA from biological samples such as tissue, blood, and cells. Proteinase K works optimally under conditions that break down proteins, facilitating the extraction of pure nucleic acids. The enzyme is essential in numerous molecular biology techniques, including PCR, cloning, and sequencing. This product is offered as a 1g vial, ensuring ample supply for multiple extractions.
Catalog No. 50-006-59998 Supplier Alkali Scientific Supplier No. OTE004CP
Proteinase K is a highly active protease enzyme used primarily in DNA extraction protocols. This enzyme efficiently digests proteins and helps in the removal of contaminants that may inhibit downstream applications. It is particularly useful in isolating high-quality DNA from biological samples such as tissue, blood, and cells. Proteinase K works optimally under conditions that break down proteins, facilitating the extraction of pure nucleic acids. The enzyme is essential in numerous molecular biology techniques, including PCR, cloning, and sequencing. This product is offered as a 1g vial, ensuring ample supply for multiple extractions.