Learn More
Alkali Scientific Proteinase K, 100 Mg
SDP
Catalog No. 5000660010
Encompass_Preferred
Proteinase K is a high-activity protease enzyme used primarily in DNA and RNA extraction procedures. This enzyme is highly effective in breaking down proteins, including histones, which helps release nucleic acids from cellular and nuclear structures. Proteinase K remains active under a wide range of conditions, making it ideal for use in DNA extractions that involve detergents or other chemical treatments. This reagent is provided in a 100 mg package, suitable for larger experiments or multiple applications. It is frequently used in research labs, particularly in protocols for DNA purification, proteinase inactivation, and enzymatic assays.
Catalog No. 50-006-60010 Supplier Alkali Scientific Supplier No. OTE002CP
Proteinase K is a high-activity protease enzyme used primarily in DNA and RNA extraction procedures. This enzyme is highly effective in breaking down proteins, including histones, which helps release nucleic acids from cellular and nuclear structures. Proteinase K remains active under a wide range of conditions, making it ideal for use in DNA extractions that involve detergents or other chemical treatments. This reagent is provided in a 100 mg package, suitable for larger experiments or multiple applications. It is frequently used in research labs, particularly in protocols for DNA purification, proteinase inactivation, and enzymatic assays.