Learn More
Abcam Proteinase K Antigen Retrieval Solution, 4ML
Catalog No. NC1239312
Encompass_Preferred
Proteinase K antigen retrieval solution is a ready-to-use, single-component solution designed for proteolytic-induced antigen retrieval (PIER) in immunohistochemistry (IHC) and pre-treatment for in situ hybridization (ISH). - Cited in over 25 publications - Ready-to-Use – no preparation needed for immediate application - Incubate tissue section for 5-7 minutes at room temperature in IHC - Incubate tissue section for 15 minutes at 37°C in ISH
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1239312 Supplier Abcam Supplier No. AB642204ML
Proteinase K antigen retrieval solution is a ready-to-use, single-component solution designed for proteolytic-induced antigen retrieval (PIER) in immunohistochemistry (IHC) and pre-treatment for in situ hybridization (ISH). - Cited in over 25 publications - Ready-to-Use – no preparation needed for immediate application - Incubate tissue section for 5-7 minutes at room temperature in IHC - Incubate tissue section for 15 minutes at 37°C in ISH
The product is subject to the following: Abcam Restricted Use Statement