Learn More
Takara Bio Proteinase K, 100 mg
SDP
Catalog No. NC1288588
Encompass
Proteinase K is a robust endopeptidase that inactivates and destroys proteins without any pronounced cleavage specificity. Consists of 100 mg of lyophilized Proteinase K (with an activity of >2.5 U/mg) plus 8 ml of a stabilizing solution (Proteinase Buffer PB).
Catalog No. NC1288588 Supplier Takara Bio Supplier No. 740506
Proteinase K is a robust endopeptidase that inactivates and destroys proteins without any pronounced cleavage specificity. Consists of 100 mg of lyophilized Proteinase K (with an activity of >2.5 U/mg) plus 8 ml of a stabilizing solution (Proteinase Buffer PB).