Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Omega Biotek Inc Proteinase K, 10 mL solution, 20 mg/mL concentration, non-specific serine protease, RNase/DNase free, high stability in SDS, EDTA, urea

Catalog No. NC1926795
Encompass

Omega Bio-tek Proteinase K is a non-specific serine protease that is essential for various molecular biology applications. Supplied as a ready-to-use 10 mL solution, it effectively inactivates nucleases and promotes cell lysis, ensuring high-quality DNA and RNA isolation. This enzyme is stable across a broad pH range and maintains its activity even in the presence of protein-denaturing agents such as SDS, EDTA, and urea.

  • Volume: 10 ml
  • Formulation: Ready-to-use solution
  • Concentration: 20 mg/ml
  • Activity: >600 mAU/mg
  • Enzyme type: Non-specific serine protease
  • Source organism: Tritirachium album
  • Stability: High stability in SDS, EDTA, urea
  • pH range: Wide range
  • Primary application: Inactivation of nucleases and lysis for DNA/RNA isolation
  • Purity: RNase/DNase free
  • Storage temperature: -20°C
  • Shipping condition: Requires ice pack
  • Country of origin: USA

Catalog No. NC1926795 Supplier Omega Biotek Inc Supplier No. AC116
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Omega Bio-tek Proteinase K is a non-specific serine protease that is essential for various molecular biology applications. Supplied as a ready-to-use 10 mL solution, it effectively inactivates nucleases and promotes cell lysis, ensuring high-quality DNA and RNA isolation. This enzyme is stable across a broad pH range and maintains its activity even in the presence of protein-denaturing agents such as SDS, EDTA, and urea.

  • Volume: 10 ml
  • Formulation: Ready-to-use solution
  • Concentration: 20 mg/ml
  • Activity: >600 mAU/mg
  • Enzyme type: Non-specific serine protease
  • Source organism: Tritirachium album
  • Stability: High stability in SDS, EDTA, urea
  • pH range: Wide range
  • Primary application: Inactivation of nucleases and lysis for DNA/RNA isolation
  • Purity: RNase/DNase free
  • Storage temperature: -20°C
  • Shipping condition: Requires ice pack
  • Country of origin: USA

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.