Learn More
Diagnostic Biosystems PROTEINASE K 25X Proteinase K
Catalog No. 502407464
Encompass_Preferred
Proteinase K is specifically formulated to achieve proteolytic digestion of formalin fixed, paraffin embedded tissue sections prior to performing in situ hybridization procedures.\n\nProteolytic digestion of formalin fixed tissues improves the accessibility of target DNA to biotinylated probes, which results in better in situ staining.
Catalog No. 50-240-7464 Supplier Diagnostic Biosystems Supplier No. K030
Proteinase K is specifically formulated to achieve proteolytic digestion of formalin fixed, paraffin embedded tissue sections prior to performing in situ hybridization procedures.\n\nProteolytic digestion of formalin fixed tissues improves the accessibility of target DNA to biotinylated probes, which results in better in situ staining.