Learn More
Enzo Life Sciences Proteinase K (5 mg) CAS: 39450-01-6
SDP
Catalog No. NC1704678
Encompass_Preferred
PCR grade proteinase K used for molecular biology applications. ENZO Proteinase K is specifically designed for use in the pretreatment of formalin-fixed tissue and paraformaldehyde-fixed cells in preparation for in situ hybridization and detection studies. Formulation: Lyophilized. Source: Pichia pastoris UniProt: P06873. Long Term Storage: +4°C
Catalog No. NC1704678 Supplier Enzo Life Sciences Supplier No. ENZ33801
PCR grade proteinase K used for molecular biology applications. ENZO Proteinase K is specifically designed for use in the pretreatment of formalin-fixed tissue and paraformaldehyde-fixed cells in preparation for in situ hybridization and detection studies. Formulation: Lyophilized. Source: Pichia pastoris UniProt: P06873. Long Term Storage: +4°C