Learn More
Agilent Technologies PROTEINASE K
Catalog No. 50125520
Encompass
Proteinase K, Broad spectrum protease, digestion of protein in biological samples, sp.activity 20 U/microgram at 37degreeC, Preparing chromosomal DNA for pulse field electrophoresis, protein fingerprinting, Nuclease removal from DNA and RNA
Catalog No. 50-125-520 Supplier Agilent Technologies Supplier No. 300140
Proteinase K, Broad spectrum protease, digestion of protein in biological samples, sp.activity 20 U/microgram at 37degreeC, Preparing chromosomal DNA for pulse field electrophoresis, protein fingerprinting, Nuclease removal from DNA and RNA