Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Proteinase K (50 μg/μL)

Catalog No. QS0512
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
750 μL
3.5 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QS0512 3.5 mL
QS0511 750 μL
QS0510 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. QS0512 Supplier Invitrogen™ Supplier No. QS0512
Add to Cart
Add to Cart

This Proteinase K is used in QuantiGene assays and is one of two components in the QuantiGene Sample Processing kits.

Proteinase K digests proteins such as ribonucleases that could interfere with the quantitation of RNA using QuantiGene assays.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Shipping Condition Blue Ice
Form Liquid
Quantity 3.5 mL
Product Type Proteinase K

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.