Learn More
Alkali Scientific Proteinase K, 50 Milligrams
SDP
Catalog No. 5000660006
Encompass_Preferred
Proteinase K is a broad-spectrum serine protease widely used in molecular biology for the digestion of proteins and removal of enzymatic contaminants. It is particularly effective in breaking down proteins during DNA and RNA extraction procedures, ensuring high purity of nucleic acids. The enzyme remains active under a wide range of temperatures and pH conditions and is capable of digesting proteins even in the presence of detergents and denaturing agents. Proteinase K is commonly used to inactivate nucleases, degrade structural proteins, and improve yield during sample preparation. It is an essential reagent in genomics, forensic analysis, and diagnostic workflows requiring efficient protein removal.
Catalog No. 50-006-60006 Supplier Alkali Scientific Supplier No. OTE005CP
Proteinase K is a broad-spectrum serine protease widely used in molecular biology for the digestion of proteins and removal of enzymatic contaminants. It is particularly effective in breaking down proteins during DNA and RNA extraction procedures, ensuring high purity of nucleic acids. The enzyme remains active under a wide range of temperatures and pH conditions and is capable of digesting proteins even in the presence of detergents and denaturing agents. Proteinase K is commonly used to inactivate nucleases, degrade structural proteins, and improve yield during sample preparation. It is an essential reagent in genomics, forensic analysis, and diagnostic workflows requiring efficient protein removal.