Learn More
AG Scientific Inc Proteinase K, 500 MG
Supplier:  AG Scientific Inc P1264500MG
Encompass_Preferred
Proteinase K is a highly reactive serine protease that displays an ability to digest native proteins, thereby inactivating enzymes such as DNase and RNase without recourse to a denaturation process. It is the most powerful proteinase among all proteinases characterized so far. It cleaves at the peptide bond adjacent to the carboxylic acid group of aliphatic, aromatic or hydrophobic amino acids. Recombinant Proteinase K is used in the isolation or preparation of high molecular weight nucleic acids. It is highly pure and has a higher specific activity and is more stable at room temperature when compared to native Proteinase K.CAS Number: 39450-01-06Molecular Weight: 29.3 kDaSolubility: 50mM Tris-HCl (pH 7.5), 3mM CaCl2, 50% GlycerolStorage Temperature: +4CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-6964