Learn More
Alkali Scientific Proteinase K, 500 Mg
SDP
Catalog No. 5000661368
Encompass_Preferred
Proteinase K is a highly effective protease enzyme used for breaking down proteins in DNA and RNA extraction protocols. With broad specificity and high activity, Proteinase K efficiently digests proteins such as histones, which helps free nucleic acids from cellular and nuclear compartments. This enzyme is highly stable and remains active in the presence of detergents and other agents commonly used in cell lysis and protein denaturation. Proteinase K is available in a 500 mg package, providing ample material for large-scale DNA extraction and purification experiments. It is a critical reagent in molecular biology, especially in studies requiring clean and efficient nucleic acid isolation.
Catalog No. 50-006-61368 Supplier Alkali Scientific Supplier No. OTE003CP
Proteinase K is a highly effective protease enzyme used for breaking down proteins in DNA and RNA extraction protocols. With broad specificity and high activity, Proteinase K efficiently digests proteins such as histones, which helps free nucleic acids from cellular and nuclear compartments. This enzyme is highly stable and remains active in the presence of detergents and other agents commonly used in cell lysis and protein denaturation. Proteinase K is available in a 500 mg package, providing ample material for large-scale DNA extraction and purification experiments. It is a critical reagent in molecular biology, especially in studies requiring clean and efficient nucleic acid isolation.