Learn More
TARGETMOL CHEMICALS INC PROTEINASE K 500MG
SDP
Catalog No. 502579036
Encompass_Preferred
Also available in 100 mg and bulk. Please contact Fisher for quotes. Proteinase K (proteinase K for tritirachium album) is a nonspecific serine protease that is useful for general digestion of proteins. It is active in the presence of SDS or urea and over a wide range of pH (4-12) salt concentrations and temperatures. purity: 98%
Catalog No. 50-257-9036 Supplier TARGETMOL CHEMICALS INC Supplier No. T8936500MG
Also available in 100 mg and bulk. Please contact Fisher for quotes. Proteinase K (proteinase K for tritirachium album) is a nonspecific serine protease that is useful for general digestion of proteins. It is active in the presence of SDS or urea and over a wide range of pH (4-12) salt concentrations and temperatures. purity: 98%