Learn More
Sigma Aldrich Fine Chemicals Biosciences Proteinase K from Tritirachium album, 39450-01-6, MFCD00132129, 25 mg
Catalog No. NC1023919
Encompass
Lyophilized powder, BioUltra, ≥30 units/mg protein, for molecular biology, Synonym: Endopeptidase K. Useful for the proteolytic inactivation of nucleases during the isolation of DNA and RNA. Removes endotoxins that bind to cationic proteins such as lysozyme and ribonuclease A.
Catalog No. NC1023919 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P230825MG
Lyophilized powder, BioUltra, ≥30 units/mg protein, for molecular biology, Synonym: Endopeptidase K. Useful for the proteolytic inactivation of nucleases during the isolation of DNA and RNA. Removes endotoxins that bind to cationic proteins such as lysozyme and ribonuclease A.