Learn More
Sigma Aldrich Fine Chemicals Biosciences Proteinase K from Tritirac1G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences SAE00091G
Encompass
Proteinase K (PK) from fungi Tritirachium album encodes a 40 kDa protein. The N-terminal propeptide region shows homology with bacterial subtilisin. PK crystallizes even under microgravity conditions on the space shuttle mission. PK is an effective protein system for studying protein engineering process.
Catalog No. 50-290-7952