Learn More
Crescent Chemical Co Inc PROTEINASE K FROM TRITIRACHIUM
SDP
Supplier:  Crescent Chemical Co Inc SE33752.03
Encompass_Preferred
Proteinase K from Tritirachium album min. 8 DMC-U/mg lyophil. Serin protease with very broad range of action: cleaves peptide bonds at the carboxylic side of aliphatic, aromatic, and hydrophobic amino acids. Suitable for the isolation of DNA and RNA (1, 3). Size - 500MG Storage Conditions -
Catalog No. 50-248-067