Learn More
LAMDA BIOTECH INC Proteinase K Powder
Catalog No. NC3726424
Encompass_Preferred
A recombinant serine protease designed for protein removal in nucleic acid preparation protocols. This enzyme is produced through recombinant expression for molecular biology applications.
Catalog No. NC3726424 Supplier LAMDA BIOTECH INC Supplier No. DB04523
A recombinant serine protease designed for protein removal in nucleic acid preparation protocols. This enzyme is produced through recombinant expression for molecular biology applications.