Learn More
Sigma Aldrich Fine Chemicals Biosciences Proteinase K, recombinant, PCR Grade, 5 mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 3115828001
Encompass
This PCR grade Proteinase K is extremely effective on native proteins and can therefore be used to rapidly inactivate endogenous nucleases such as RNases and DNases. This property makes Proteinase K particularly suitable for the isolation of native RNA and DNA from tissues or cell lines.
Catalog No. NC1145418