Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Proteinase K Recombinant Protein

Catalog No. PIRP87705
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP87705 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP87705 Supplier Invitrogen™ Supplier No. RP87705
Add to Cart
Edge
Add to Cart

Recombinant Protein

Animal Origin Free No DNase contamination detected by digestion of lambda DNA at 37°C for 6 hours. No RNases contamination detected by digestion of lambda DNA at 25°C for 16 hours. Host genomic DNA/RNA is no residual detected by PCR. Storage: Store the lyophilized protein at 4°C for up to 1 year and less than -20°C for long term. Lyophilized powder is recommended to dissolve in in 20mM Tris-Hcl, 30mM NaCl, 50% Glycerol, 5mM CaCl2, pH 7.2-7.4. After reconstitution, the protein solution is stable at -20°C for 1 year, at 2-8°C for up to 6 months.

Proteinase K (E.C.3.4.21.64) is an extracellular serine endoproteinase produced by Tritirachinm album Limber. Proteinase K is active in a wide range of temperatures and buffers with optimal activity between 20 and 6°C and a pH between 7.5 and 12.0. Activity is stimulated when up to 2% SDS or up to 4 M urea are included in the reaction. Calcium is important for thermostability of Proteinase K but it is not required for catalysis, therefore Proteinase K is also active in buffers containing chelating agents such as EDTA.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Control
Formulation protein with no preservative; pH 7.2-7.4
Name Proteinase K
Quantity 1 g
Source Yeast
Gene Alias EC 3.4.21.14; Protease K
Common Name Proteinase K
Biological Activity Measured by its digestion activity. The activity is >30 U/mg One unit will digest Casein (at 37°C (pH 7.5) / minute to produce equal absorbance as 1.0 μmol of L-tyrosine.)
Conjugate Unconjugated
Recombinant Recombinant
Sequence Full length Proteinase K protein
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Expression System P. pastoris
Form Lyophilized
Purity or Quality Grade >95% by SDS-Page
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.