Learn More
Gold Biotechnology Inc Proteinase K, RNase/DNase free 5 x 1 g
SDP
Catalog No. NC1391855
Encompass
Proteinase K is a highly reactive nonspecific serine protease that belongs to the subtilisin family of proteins. It cleaves at the carboxylic acid side of aliphatic, aromatic, or hydrophobic amino acids. Proteinase K is capable of inactivating RNases and DNases and is used in the isolation or preparation of high molecular weight nucleic acids. Proteinase K is also useful for helping to characterize enzymes, due to its cleavage specificity. This enzyme was designated proteinase K because of its ability to hydrolyze keratin. Proteinase K is stable in a wide variety of detergents and buffer salts and at various temperatures and pH. The isoelectric point of proteinase K is 8.9.
Catalog No. NC1391855 Supplier Gold Biotechnology Inc Supplier No. P4805
Proteinase K is a highly reactive nonspecific serine protease that belongs to the subtilisin family of proteins. It cleaves at the carboxylic acid side of aliphatic, aromatic, or hydrophobic amino acids. Proteinase K is capable of inactivating RNases and DNases and is used in the isolation or preparation of high molecular weight nucleic acids. Proteinase K is also useful for helping to characterize enzymes, due to its cleavage specificity. This enzyme was designated proteinase K because of its ability to hydrolyze keratin. Proteinase K is stable in a wide variety of detergents and buffer salts and at various temperatures and pH. The isoelectric point of proteinase K is 8.9.