Learn More
Gold Biotechnology Inc Proteinase K Solution 20 mg/ml 2 mL
SDP
Catalog No. NC1373871
Encompass_Preferred
Proteinase K is a highly reactive nonspecific serine protease that belongs to the subtilisin family of proteins. It cleaves at the carboxylic acid side of aliphatic, aromatic, or hydrophobic amino acids. Proteinase K is capable of inactivating RNases and DNases and is used in the isolation or preparation of high molecular weight nucleic acids. Proteinase K is also useful for helping to characterize enzymes, due to its cleavage specificity. This enzyme was designated proteinase K because of its ability to hydrolyze keratin. Proteinase K is stable in a wide variety of detergents and buffer salts and at various temperatures and pH. The isoelectric point of proteinase K is 8.9.
Catalog No. NC1373871 Supplier Gold Biotechnology Inc Supplier No. P480SL2
Proteinase K is a highly reactive nonspecific serine protease that belongs to the subtilisin family of proteins. It cleaves at the carboxylic acid side of aliphatic, aromatic, or hydrophobic amino acids. Proteinase K is capable of inactivating RNases and DNases and is used in the isolation or preparation of high molecular weight nucleic acids. Proteinase K is also useful for helping to characterize enzymes, due to its cleavage specificity. This enzyme was designated proteinase K because of its ability to hydrolyze keratin. Proteinase K is stable in a wide variety of detergents and buffer salts and at various temperatures and pH. The isoelectric point of proteinase K is 8.9.